Supelco® Ascentis® Express F5 (PFP) HPLC and UHPLC Columns
Supplier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
53566-U 53567-U 53568-U 53569-U 53571-U 53574-U 53576-U 53577-U 53578-U 53579-U 53581-U 53583-U 53584-U 53590-U 53591-U 53592-U 53594-U 53597-U 53599-U 50603-U 50604-U 50605-U 50607-U 50612-U 50613-U 50614-U 50615-U 50616-U 50619-U 50622-U 50623-U 50624-U 50625-U 50626-U 50627-U 50628-U 50631-U 50632-U 50633-U 50634-U 50635-U 50857-U 50858-U 50859-U 50861-U 50863-U 50867-U 50869-U 50871-U 50876-U 50879-U 50881-U 50884-U 50886-U
53566-UEA 428 GBP
53566-U 53567-U 53568-U 53569-U 53571-U 53574-U 53576-U 53577-U 53578-U 53579-U 53581-U 53583-U 53584-U 53590-U 53591-U 53592-U 53594-U 53597-U 53599-U 50603-U 50604-U 50605-U 50607-U 50612-U 50613-U 50614-U 50615-U 50616-U 50619-U 50622-U 50623-U 50624-U 50625-U 50626-U 50627-U 50628-U 50631-U 50632-U 50633-U 50634-U 50635-U 50857-U 50858-U 50859-U 50861-U 50863-U 50867-U 50869-U 50871-U 50876-U 50879-U 50881-U 50884-U 50886-U
Supelco® Ascentis® Express F5 (PFP) HPLC and UHPLC Columns
Chromatography Columns
Ascentis® Express F5 (PFP) high-speed, high-performance liquid chromatography columns are based on the highly efficient Fused-Core® particle design.
This particle design exhibits very high column efficiency due to the shallow diffusion paths in the 0,4 μm thick porous shell and the small overall particle size. The bonded, endcapped, dimethylpentafluorophenylpropyl stationary phase of Ascentis® Express F5 provides a stable, reversed phase packing with electron-deficient phenyl rings due to the presence of electronegative fluorines. In addition to forming pi-pi and mildly stericMore Product Information