Potassium hydroxide 0.23 mol/l (0.23 N) in aqueous solution, AVS TITRINORM volumetric solution

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Potash caustic, Caustic potash

5045.9010EA 85.2 GBP
5045.9010
Potassium hydroxide 0.23 mol/l (0.23 N) in aqueous solution, AVS TITRINORM volumetric solution
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Density: 1.009 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 0
UN: 1814
ADR: 8 III
Merck Index: 12,07806

Order Now

Specification Test Results

Titer 0,2185 - 0,2415 mol/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR