Avantor® Apex ODS, HPLC Columns
Supplier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
4M15310EA 877 GBP
4M15310 4M3310P 4M5310 4M10310 4M25310 FH1310-2/N
Avantor® Apex ODS, HPLC Columns
Chromatography Columns
Avantor® Apex and Avantor® Apex II HPLC column phases are recommended for routine analyses.
  • Conventional 100 Å pore size spherical silica
  • Narrow particle distribution
  • Controlled surface area
  • USP L1