J.T.Baker®, Robotic Tips Type DX
Supplier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
49136-0000EA 47.4 GBP
49136-0000 49137-0000 49136-2000 49137-2000
J.T.Baker®, Robotic Tips Type DX
Pipette Tips
Suitable for use on Dynex DSX and DS2 instruments. Available in non conductive version, without filter, in container rack, and in CE-IVD marked version for clinical diagnostic applications.
  • 108 tips in one optimized container with lid
  • Especially developed for the automated sample handling
  • Highest quality specifications for the compatibility and cleanliness
  • Blue sample tips for visual tip identification
The validation of the tips on the instrument for specific applications is within the responsibility of the user.