D-(+)-Glucose, anhydrous, granular, AR® ACS, Macron Fine Chemicals™

Supplier: Avantor Performance Materials
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose

4912-06EA 0 GBP
4912-06 4912-07 4912-20 4912-24
D-(+)-Glucose, anhydrous, granular, AR® ACS, Macron Fine Chemicals™
D(+)-Glucose

Protect and stabilise valuable biochemical proteins and streamline downstream processing with reliable, high-performance sugars.

Formula: C₆H₁₂O₆
Melting Pt: 146 °C
Density: 1.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00148912
CAS Number: 50-99-7
EINECS: 200-075-1
Merck Index: 14,04459
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ec28c7fd4803097405f8eaef3171b3a40d40c5&vid=62627c96e2ae9f2a0cae732d0ddf0a100247298e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements ≤0.01%
For Laboratory,Research,or Manufacturing Use ≤5 ppm
ACS - Chloride (Cl) ≤0.01 %
ACS - Heavy Metals (as Pb) ≤5 ppm
ACS - Insoluble Matter ≤0.005 %
ACS - Iron (Fe) ≤5 ppm
ACS - Loss on Drying at 105°C ≤0.2 %
ACS - Residue after Ignition ≤0.02 %
ACS - Starch Passes Test
ACS - Sulfate and Sulfite (as SO₄) ≤0.005 %
ACS - Specific Rotation [Å]²⁵^D (+) 52.5 - 53.0 Degree
ACS - Titrable Acid (meq/g) ≤0.002


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR