Autoclaving baskets, Nalgene®
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
6917-0127 6917-0150 6917-0230
489-2150EA 48.2 GBP
489-2150 489-2151 489-2152
Autoclaving baskets, Nalgene®
Baskets Autoclavable Baskets
PP.
  • Smooth surfaces will not scratch plastic or glass
  • Provides airflow for venting and drying
  • Three sizes available
  • Nestable for storage
These autoclavable baskets are designed for holding labware during autoclaving and washing.
Colour: White