Sterilising pans, Nalgene®
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
6900-0010 6900-0020
489-2120EA 45.6 GBP
489-2120 489-2121
Sterilising pans, Nalgene®
Trays/Dishes/Pots Sterilising Trays/Pans
PP.
  • Rolled edges for extra stiffness
  • Handles for easy carrying
For drying and sterilising instruments and labware, collecting and washing soiled lab equipment.