MES sodium salt ≥85.0% for biopharmaceutical production, purified, J.T.Baker®

Supplier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

4813-02EA 2300 GBP
4813-02 4813-06 4813-01
MES sodium salt ≥85.0% for biopharmaceutical production, purified, J.T.Baker®
MES sodium salt

Suitable for use in biopharmaceutical manufacturing applications.

Formula: C₆H₁₂NNaO₄S
MDL Number: MFCD00065473
CAS Number: 71119-23-8
EINECS: 275-203-2

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use ≥85.0%
Assay (C₆H₁₂NNaO₄S) ≥85.0 %
Appearance Passes Test
Identification (by IR) Passes Test
pH of 1% Aqueous Solution at 25°C 8.9 - 10.1
Solubility Passes Test
Ultraviolet Absorbance of a 10% Aqueous Solution - 280 nm ≤0.03
Ultraviolet Absorbance of a 10% Aqueous Solution - 260 nm ≤0.05
Water (H₂O) ≤15.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR