Vaccum Pro Horizontal Laboratory Autoclaves
Supplier: CERTOCLAV
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
8500487 8500489
CERT8500487EA 6240 GBP
CERT8500487 481-0910
Vaccum Pro Horizontal Laboratory Autoclaves
Sterilisers Autoclaves (Steam Sterilisers)
These compact stand-alone laboratory autoclaves CertoClav-Vacuum-Professional offer 11 pre-defined programs for sterilisation of solid materials, hollow body, porous materials, textiles, liquids prions and plastics. They are used in microbiology, bio-chemistry, food industry, material testing, pharmacy and universities.
  • Sterilisation of liquids and solids
  • Stand-alone: No external water connection necessary
  • The pre-heat able chamber, cooling-ventilation and vacuum pump guarantee short cycle times from 20 minutes for solid materials including drying time
More Product Information