VWR® VAPOUR-Line, Vertical Autoclaves with Exhaust Air Filtration (Option FA)
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
481-0782EA 13460 GBP
481-0782 481-0783 481-0784 481-0785 481-0786 481-0787 481-0795 481-0796 481-0797 481-0798
VWR® VAPOUR-Line, Vertical Autoclaves with Exhaust Air Filtration (Option FA)
Sterilisers Autoclaves (Steam Sterilisers)
These autoclaves are ideal for media, equipment and waste sterilisation. Designed for safety level laboratories, they are equipped with exhaust air filtration (option FA). This protects users and the environment against germs and genetically-modified material which is blown out of the autoclave into the laboratory during the de-aeration phase.
  • Easy to install, operate and service
  • With or without integrated fast cooling for liquids
  • Exhaust air filtration
  • Depth filter with metal housing does not distort
  • Integrated batch counter
  • Inline sterilisation of filter
More Product Information