Autoclaves, portable,VARIO Series
Supplier: DIXONS SURGICAL INSTR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ST3028V ST2228V
681-0003EA 0 GBP
681-0003 481-0374
Autoclaves, portable,VARIO Series
Sterilisers Autoclaves (Steam Sterilisers)
This range of portable autoclaves are designed for safety and reliability, yet remain easy to use and maintain. They are fitted with an automatic cut-out to prevent them boiling dry. The lid is removable for full width access, items can either be placed in the inner liner provided or in accessory dressing drums or basket. All models are heated with a 2 kW single internal element. Temperature is controlled by the VARIO time and temperature controller and a Pt 100 thermocouple. The VARIO cycle control features four preset cycles and one variable cycle. The variable cycle is stored in permanentMore Product Information