Racks for cryogenic vials, Nalgene®
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5030-0505
479-3228EA 23.2 GBP
479-3228
Racks for cryogenic vials, Nalgene®
Racks Cryogenic Vial Racks
PC, autoclavable.
  • Bottom interlocks with base of vial
  • Spacing around each hole allows vials to be easily removed
Allows single handed vial manipulation.