VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
479-1635EA 5 GBP
479-1635 479-1392 479-1393 479-1394 479-1395 479-1396 479-1402 479-1403 479-1404 479-1405 479-1406 479-1412 479-1413 479-1414 479-1415 479-1416 479-1422 479-1423 479-1424 479-1425 479-1426 479-1432 479-1433 479-1434 479-1435 479-1436 479-1451 479-1452 479-1453 479-1454 479-1455 479-1456 479-1457 479-1458 479-1471 479-1472 479-1473 479-1474 479-1475 479-1476 479-1477 479-1478 479-1479 479-1480 479-1481 479-1482 479-1491 479-1492 479-1493 479-1494 479-1495 479-1496 479-1497 479-1498 479-1627 479-1628 479-1629 479-1630 479-1631 479-1632 479-1633 479-1634
VWR®, Cryoboxes, Cardboard, 136x136 mm
Boxes Cryoboxes
Cardboard.
  • Waterproof coating
  • Various combinations possible with different dividers (to be ordered separately)
  • Available in different heights and five colours
For storage of cryovials at low temperatures.
Freezable to –140 °C.