Storage tubes, barcoded, Corning®
Supplier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
8500 8501 8502 8503 8509 8510
479-0968EA 816 GBP
479-0968 479-0969 479-0970 479-0971 479-0972 479-0973
Storage tubes, barcoded, Corning®
Tubes Microtubes
PP. The 1D/2D storage line is an innovative product with both a linear 1D and 2D barcodes etched on each tube.
  • Maximum information, 14×14 dot 2D barcode
  • Barcode stability, laser-etched, not an attached label
  • Compound compatibility, polypropylene construction throughout, making it inert