Transport system for cooled samples, minivaqbox
Supplier: DELTA T
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
V020010 V020011 V020012
478-9925EA 1270 GBP
478-9925 478-9926 478-9927
Transport system for cooled samples, minivaqbox
Boxes Cooling/Freezing Boxes and Bags
This transport system uses vacuum insulation technology and is ideal for lengthy refrigerated transport of specimens, cells, and other sensitive items. The box is provided with a shoulder strap to make carrying easier. Use of the MiniVaqBox transport system with Delta T elements will ensure proper temperature maintenance without the application of any external energy for a period of 48 hours. First, the elements must be pre-cooled for a certain period of time until the fluid is solid. This will occur when the temperature is approximately 5 °C below the fluid’s melting point. The&More Product Information