Dewar flasks, type GSS
Supplier: KGW
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2300 2302 2303 2304 2305 2680 2681 2682 2301
478-5432EA 662 GBP
478-5432 478-5435 478-5434 478-5431 478-5436 478-5142 478-5143 478-5144 478-5433
Dewar flasks, type GSS
Dewars and Cryogenic Vessels
Dewar flasks in stainless steel for storage and transport of liquid nitrogen, dry ice and other coolants. They are vacuum insulated and have a temperature range from −200 to +200 °C.
  • With side grip and transport handle (except model GSS 500)