Locator™ Cryogenic Rack and Box Systems
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
CY50945
478-0809EA 5500 GBP
478-0809
Locator™ Cryogenic Rack and Box Systems
Racks Cryogenic Box Racks
Retrieve cell samples quickly and easily using Thermo Scientific™ Locator™ cryogenic rack and box systems, which feature a unique hanging rack and gridded box design, advanced vacuum insulation, unmatched temperature uniformity and an innovative liquid nitrogen monitor.
  • Compact design minimises space requirements while maximising storage capacity
  • System is suitable for both manual and computerised inventory record-keeping methods
  • Super-insulated vessels have special, narrow-mouth design that conserves liquid nitrogen by minimising evaporation
  • Samples are
More Product Information