3,3,5-Trimethylcyclohexyl salicylate (cis and trans mixture) 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

474510250 474511000
474510250.EA 22.9 GBP
474510250. 474511000.
3,3,5-Trimethylcyclohexyl salicylate (cis and trans mixture) 98%
3,3,5-Trimethylcyclohexyl salicylate (cis and trans mixture)
Formula: C₁₆H₂₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00019377
CAS Number: 118-56-9
EINECS: 204-260-8

Order Now

Specification Test Results

Appearance clear colorless liquid
Infrared spectrum Conforms
HPLC ≥97.5 % sum of isomers


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR