4,4'-(4,4'-Isopropylidenediphenoxy)diphthalic anhydride 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: BPADA, 4,4'-(4,4'-Isopropylidenediphenoxy)bis(phthalic anhydride), 5,5'-((Propane-2,2-diylbis(4,1-phenylene))bis(oxy))bis(isobenzofuran-1,3-dione)

474470500 474472500
474470500.EA 44 GBP
474470500. 474472500.
4,4'-(4,4'-Isopropylidenediphenoxy)diphthalic anhydride 98%
4,4'-(4,4'-Isopropylidenediphenoxy)diphthalic anhydride
Formula: C₃₁H₂₀O₈
Boiling Pt: 712°C (760mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00319167
CAS Number: 38103-06-9
EINECS: 253-781-7

Order Now

Specification Test Results

Appearance almost white to yellow powder or crystals
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR