Ice machine, flakes
Supplier: PORKKA SCANDINAVIA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
KF165 KF75 KF45 KF145 KF185 KF85
474-1635EA 4760 GBP
474-1635 472-0385 474-0102 474-0100 474-0101 474-0103
Ice machine, flakes
Ice Makers
Ice flakers produce ice flakes that are dry and compact, this allows the ice to remain free flowing for easy use even after long storage periods.They have been manufactured to the highest specification, this includes a body work of high quality stainless steel grade 304 S 01. This eliminates any possibility of rusting when working in harsh environments. All internal components utilised are of either ABS (food grade) plastic or materials for use with potable water.
  • Used in many diverse areas: food, supermarket, laboratories, hopitals...
  • Air cooled
  • Legs as
More Product Information