Freezers
Supplier: GRAM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MIDIF425LSH MIDIF625LSH PLUSF600RSH PLUSF660RSH PLUSF1270RSH PLUSF1400RSH COMPACTF210LG COMPACTF410LG COMPACTF610LG
471-0547EA 0 GBP
471-0547 471-0548 471-0534 471-0535 471-0533 471-0826 471-0828 471-0829 471-0830
Freezers
Freezers Laboratory / General Purpose Freezers
The unique air distribution system combined with a finned tube evaporator results in excellent temperature consistency for biostorage: the evaporator design eliminates the risk of 'cold walls', which can damage delicate items stored touching the sides of the cabinet. Whilst the air distribution system which directs the cold air down the rear of the cabinet and back up to the evaporator fan at the top, ensures a uniform temperature throughout.
Freezers Compact
The Compact range cabinets are general purpose commercial freezers for basic biostorage purposes where the focus is on reliabilityMore Product Information