ATEX Freezers, Bioline range
Supplier: GRAM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BF600DLG BR1270LG BR600DLG RF1270LG RF425LG RF600DLG RF625LG RF660DLG BF1270LG IIRF210LG IIRF410LG IIRF610LG EF425LG
471-0521EA 0 GBP
471-0521 471-0522 471-0523 471-0505 471-0509 471-0506 471-0510 471-0507 471-0602 471-0807 471-0808 471-0809 471-0842
ATEX Freezers, Bioline range
Freezers Laboratory / General Purpose Freezers
BioLine is a comprehensive refrigeration range designed to deal with special bioscience needs. BioLine is distinct from other 'spark-free' freezers on the market, as both the interior and the exterior of all units have been shown to comply with the European guidelines contained in the ATEX [EN/IEC60079-15] directive which means they are safe for use in potentially explosive atmospheres (Zone 2 [Category 3]). The unique air distribution system is combined with BioLine’s innovative 'smart defrost' function to maintain temperature consistency for biostorage: Smart defrost is an intelligentMore Product Information