Thiazolidine-4-carboxylic acid 95%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

470800010 470800050
470800010.EA 52.1 GBP
470800010. 470800050.
Thiazolidine-4-carboxylic acid 95%
Thiazolidine-4-carboxylic acid
Formula: C₄H₇NO₂S
Storage Temperature: Ambient
MDL Number: MFCD00128958
CAS Number: 444-27-9
EINECS: 207-146-6

Order Now

Specification Test Results

Appearance white to yellow to brown powder or crystals
Infrared spectrum Conforms
HPLC ≥94 %
Titration with NaOH 94 to 106 % (On dry substance)
Water ≤10 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR