Ethanol ∼40%, technical (denatured with 5% methanol)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

466480050
466480050.EA 102 GBP
466480050.
Ethanol ∼40%, technical (denatured with 5% methanol)
Ethanol
Formula: C₂H₅OH
Density: 0.95
Flash Pt: 26°C
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Appearance clear colorless liquid
Density (20°C) 0.943 to 0.953 kg/l
Ethanol 37 to 43 v/v % (calculated)
Additional info Denatured with 5% methanol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR