Drying cabinets, insulated, E3
Supplier: GENLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
E3DWC100/N E3DWC100/F E3DWC200/F E3DWC200/N E3DWC425/F E3DWC425/N E3DWC885/F/TDI
466-0459EA 1610 GBP
466-0459 466-0458 466-0460 466-0461 466-0462 466-0464 466-0467
Drying cabinets, insulated, E3
Ovens Drying Cabinets, Glassware
These Genlab E3 drying cabinets are suitable for general purpose warming applications and drying glassware or instruments. All models in this range are fully insulated for improved efficiency and energy savings.
  • Fully insulated for improved efficiency and energy savings
  • Integral 7 day timer with time display
  • Digital temperature control with fixed over-temperature cut-out
  • An adjustable vent cover is fitted to allow further reduction of heat loss when used for warming processes
  • Controls and fan (/F models only) in base
Made from sheet steel finished inMore Product Information