Dibromo(1,5-cyclooctadiene)palladium(II) (28,4% Pd)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Dibromo(1,5-cyclooctadiene)palladium

46492.03
46492.03EA 0 GBP
46492.03
Dibromo(1,5-cyclooctadiene)palladium(II) (28,4% Pd)
Dibromo(1,5-cyclooctadiene)palladium(II)

CAS No.: 12145-47-0 Molecular Formula: C8H12Br2Pd Formula Weight: 374.41 Physical Form: Powder Appearance: Orange Melting Point: 201-205° MDL No.: MFCD00799635

Formula: C₈H₁₂Br₂Pd
MW: 374.412 g/mol
Melting Pt: 201…205 °C
Storage Temperature: Ambient
MDL Number: MFCD00799635
CAS Number: 12145-47-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR