Graphite ≥99%, powder APS 7-11 µm

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

46304.A1 46304.14
46304.A1EA 63.6 GBP
46304.A1 46304.14
Graphite ≥99%, powder APS 7-11 µm
Graphite

Purity: 99% CAS No.: 7782-42-5 Odour: Odourless UN No.: 1325 MDL No.: MFCD00144065

Formula: C
MW: 12.01 g/mol
Melting Pt: 3827
Density: 2.09…2.23 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00144065
CAS Number: 7782-42-5
EINECS: 231-95593
UN: 1325
ADR: 4.1 III
Merck Index: 12,04560

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR