Accessories for JB Academy, JB Nova, SUB Aqua Pro, SUB Aqua Plus, JB Aqua Plus and SBB Aqua Plus baths
Supplier: GRANT INSTRUMENTS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
LF14 LF28 LF6 LU14 LU28 LU36 LU6 RS14H RS28H RS36H SY1 J2-10 J2-13 J2-16 J2-19 J2-25 J2-30 J2-SE J2-LE AQBT26 AQBT5 AQL5 AQBT12 RS18H
462-8221EA 338 GBP
462-8221 462-8222 462-8220 462-8211 462-8212 462-8213 462-8210 462-8231 462-8232 462-8233 462-8354 462-8451 462-8452 462-8453 462-8454 462-8455 462-8456 462-8457 462-8458 GRANAQBT26 GRANAQBT5 GRANAQL5 GRANAQBT12 462-0187
Accessories for JB Academy, JB Nova, SUB Aqua Pro, SUB Aqua Plus, JB Aqua Plus and SBB Aqua Plus baths
Baths Water Baths, Unstirred
A range of different stainless steel accessories for use with JB Academy, JB Nova, SUB Aqua Pro, (also previous models: SUB Aqua Plus, JB Aqua Plus and SBB Aqua Plus baths).
  • Flat lids with ring sets allow vessels of various sizes and heights to be used in the bath
  • Sloping lids, with insulated handle, direct condensation down the side of the bath, protecting vessels in the bath
  • Clear PC lids are supplied as standard with JB Nova and SUB Aqua Pro baths
  • Raised half shelves help accommodate different sizes of vessel simultaneously by altering the effective
More Product Information