VWR®, Tube Racks for VWB Series Baths
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
462-0253EA 114 GBP
462-0253 462-0254 462-0255 462-0256 462-0257 462-0258 462-0252
VWR®, Tube Racks for VWB Series Baths
Racks Tube Racks
Tube racks suitable for: VWB 6, VWB 12, VWB 18, VWB 26, maximum 1, 4, 7, 7 racks per bath respectively.