VWR®, Heating Circulators
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
462-0219EA 4290 GBP
462-0219 462-0218 462-0217 462-0216 462-0215 462-0222 462-0220 462-0221
VWR®, Heating Circulators
Circulating Thermostats Open Circulator Systems Baths with Integral Circulators
Stainless steel circulating baths with choice of Advanced Programmable or Advanced Digital temperature controller. Both are easy to navigate with large, intuitive displays and multiple communication options including USB-A and B, RS232/485, Ethernet and external temperature probe. The reservoir drain can be accessed by removing the front panel. All models feature user-adjustable high temperature safety cut-off points as well as over-temperature protection. The DuraTop™ surface is cool to the touch even when operating at high temperatures.
  • Working temperatures from ambient 10 to 200 °C
More Product Information