Baths, unstirred, Clifton, NE series
Supplier: NICKEL ELECTRO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
NE1-28 NE2-14D NE1-4 NE1-8 NE1-14 NE1-22 NE2-4D NE2-8D NE2-22D NE2-28D NE1-2.5 NE1-56 NE2-56D
462-0033EA 1480 GBP
462-0033 462-0036 462-0034 462-0035 462-0031 462-0032 462-0040 462-0041 462-0037 462-0038 462-0123 462-0124 462-0125
Baths, unstirred, Clifton, NE series
Baths Water Baths, Unstirred
Unstirred baths with a corrosion resistant stainless steel tank and durable chemical resistant outer case.
  • Include 'low' height removable stainless steel shelf to increase working volume
  • Digital models have PID controller with LED display of actual temperature with 0,1 °C resolution
  • Built-in timer and auto setting alarms for protection on digital units
  • Control options either temperature control only or temperature and time, settings retained in memory
  • Anti-bacterial paint finish that actively inhibits bacterial growth
The heater element and sensor areMore Product Information