Bath fluids
Supplier: LAUDA
Ratings:
Avg. Ratings: 5.0 (1 Review)
Total Ratings: 1
Avg. Ratings: out of 5
LZB107 LZB109 LZB114 LZB209 LZB309 LZB106 LZB116 LZB314 LZB120 LZB206 LZB214 LZB216 LZB220 LZB306 LZB316 LZB320 LZB121 LZB221 LZB321 LZB126 LZB118 LZB218 LZB318
461-2845EA 265 GBP
461-2845 461-2851 461-2848 461-2864 461-2876 461-2849 461-2850 461-2855 461-2858 461-2861 461-2866 461-2867 461-2870 461-2873 461-2878 461-2881 461-2883 461-2884 461-2885 461-1115 461-1114 461-1116 461-1117
Bath fluids
Circulator Bath Fluids
Allow highly accurate thermostating, even at extreme temperatures.
  • Simple and safe handling, suitable for long term operation
  • Optimise thermostat life
* Denotes Silicone oil
** Only suitable for closed circuits