Refrigerated and heating circulators, MAGIO™ MS series
Supplier: JULABO GmbH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
9032704 9032705 9032707 9032713.N1 9032713.S1 9032714.N1 9032714.S1 9032716.N1 9032715.N1 9032717.N1 9032728.N1 9032757
461-1429EA 8597 GBP
461-1429 461-1430 461-1432 461-1433 461-1434 462-0638 462-0639 462-0640 JULA9032715.N1 JULA9032717.N1 JULA9032728.N1 JULA9032757
Refrigerated and heating circulators, MAGIO™ MS series
Circulating Thermostats Open Circulator Systems Heating/Cooling Circulators
The MAGIO™ series of cooling and heating circulators operate with a working temperature range of –50 to +300 °C and heating capacity of up to 3 kW. With their powerful pressure/suction pump the MAGIO™ units are ideal for external temperature applications. An extensive range of accessories and excellent dynamics mean that the MAGIO™ circulators can be modularly and individually adapted to applications.
  • Bright, easy to read display
  • Integrated programmer allows temperature time profiles to be run
  • Stainless steel wetted parts mean units are ideal for demanding
More Product Information