Heating circulators, MAGIO™ MS and MX series
Supplier: JULABO GmbH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
9032504 9033506 9033512 9033526
461-1423EA 4810 GBP
461-1423 461-1424 461-1425 461-1426
Heating circulators, MAGIO™ MS and MX series
Circulating Thermostats Open Circulator Systems Baths with Integral Circulators
The MAGIO™ series of heating circulators operate with a working temperature range of 20 to +300 °C and heating capacity of up to 3 kW. With their powerful pressure/suction pump the MAGIO™ units are ideal for external temperature applications. An extensive range of accessories and excellent dynamics mean that the MAGIO™ circulators can be modularly and individually adapted to applications.
  • Integrated programmer allows temperature time profiles to be run
  • Stainless steel wetted parts mean units are ideal for demanding external applications
  • Automatic shut off
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable