Refrigerated/heating circulators, CORIO™ CD series
Supplier: JULABO GmbH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
9012703 9012701 9012702 9012704 9012705 9012707 9012708 9012713.N1 9012713.S1 9012714.N1 9012714.S1 9012716.N1 9012715.N1 9012717.N1 9012701.N1 9012728.N1
461-0885EA 4915 GBP
461-0885 461-0883 461-0884 461-0886 461-0964 461-0998 461-0999 461-1455 JULA9012713.S1 461-1456 JULA9012714.S1 461-1457 JULA9012715.N1 JULA9012717.N1 JULA9012701.N1 JULA9012728.N1
Refrigerated/heating circulators, CORIO™ CD series
Circulating Thermostats Open Circulator Systems Heating/Cooling Circulators
These powerful and robust CORIO™ laboratory circulators provide the exact temperature, absolute precision and a wide working temperature range. The CORIO™ CD controller is an advanced unit with professional technology for demanding applications and can be used for temperature control of internal or external applications. The jet nozzle allows continuous adjustment of the pump stream in the system. The insulated stainless steel baths are durable and have an integrated drain tap. These heating/cooling circulators are ideal for temperature control of samples in a circulator bath or temperatureMore Product Information