Methyl acetate, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Acetic acid methyl ester

M-Clarity™ quality level = MQ100

45997-5ML-F 45997-1ML-F
45997-5ML-FEA 121 GBP
45997-5ML-F 45997-1ML-F
Methyl acetate, Supelco®
Methyl acetate
Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 56.9
Melting Pt: –98 °C
Density: 0.938 g/cm³ (20 °C)
Flash Pt: –13 °C
Storage Temperature: Ambient
MDL Number: MFCD00008711
CAS Number: 79-20-9
EINECS: 201-185-2
UN: 1231
ADR: 3 II
Merck Index: 13,06038
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=08afb932f28309b9663507ae91fa8caa84b79cfa&vid=4a8853233fbeece9b18081329d176fa8974f241d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR