Isopropyl acetate ≥99.5% (by GC), Puriss. p.a., Riedel-de Haën™

Supplier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

45960-1L 45960-250ML
45960-1LEA 149 GBP
45960-1L 45960-250ML
Isopropyl acetate ≥99.5% (by GC), Puriss. p.a., Riedel-de Haën™
Isopropyl acetate

Beilstein Number: 1740761

Formula: C₅H₁₀O₂
Boiling Pt: 85 °C
Melting Pt: –62 °C
Density: 0.8662 g/cm³ (25 °C)
Flash Pt: 2.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00008877
CAS Number: 108-21-4
EINECS: 203-561-1
UN: 1220
ADR: 3,II
Merck Index: 13,05225

Order Now

Specification Test Results

assay (GC) Min. 99 %
measurement uncertainty effective %
identity (NMR) complying
refractive index (n 25/D) 1.38 - 1.39
water (Karl Fischer) Max. 0.5 %
Aluminium (Al) Max. 0.5 ppm
Barium (Ba) Max. 0.1 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.5 ppm
Lithium (Li) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.02 ppm
Molybdenum (Mo) Max. 0.1 ppm
Sodium (Na) Max. 0.5 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.1 ppm
Strontium (Sr) Max. 0.1 ppm
Zinc (Zn) Max. 0.1 ppm
Autoignition Temperature 894 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR