VWR®, Crucibles, Stainless Steel 304
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
459-4501EA 21.4 GBP
459-4501 459-4502 459-4503
VWR®, Crucibles, Stainless Steel 304
Crucibles
These crucibles are made from stainless steel 304.
  • Highly resistant to chemicals and reagents
  • Withstand maximum temperature of 800 °C
Without lid.