Combustion boats
Supplier: MORGAN ADVANCED MATERIALS HALD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
L30/6 L30/7 L30/9
459-2213EA 40.5 GBP
459-2213 459-2214 459-2215
Combustion boats
Boats Combustion Boats
Glazed porcelain.
  • With recovery ring
  • Withstand maximum temperature 1000 °C