Incinerating dishes
Supplier: MORGAN ADVANCED MATERIALS HALD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
L33/1 L33/2 L33/4 L33C/00 L33C/00B L33C/4B L33C/7 L33D/2 L33D/3 L33D/4 L5032/1
459-2121EA 15.9 GBP
459-2121 459-2122 459-2123 459-2131 459-0038 459-2132 459-2134 459-2112 459-2113 459-2114 459-2141
Incinerating dishes
Boats Combustion Boats
Porcelain dishes, glazed inside and out, except base.
  • Withstand maximum temperature 1000 °C
  • Shallow form without spout, deep form with spout
The rectangular dishes have sloping sides.
Measurements shown are W×D×H for rectangular dishes and Ø×H for others.