VWR®, Incinerating Dishes
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
459-0233EA 41.3 GBP
459-0233 459-0232 459-0231 459-0230 459-0229 459-0228 459-0227
VWR®, Incinerating Dishes
Boats Combustion Boats
Porcelain, glazed, produced according to DIN 12904.
  • Dishes can withstand temperatures up to 1000 °C
  • Stable with large flat unglazed base
  • Glazed porcelain has high resistance to acids and bases