VWR®, Crucibles, Porcelain
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
459-0209EA 39.2 GBP
459-0209 459-0208 459-0207 459-0198 459-0197 459-0196 459-0195 459-0194 459-0201 459-0210 459-0214 459-0213 459-0199 459-0211 459-0200 459-0206 459-0205 459-0204 459-0203 459-0202 459-0212
VWR®, Crucibles, Porcelain
Crucibles
Porcelain, glazed inside and out, except base.
  • Withstands maximum temperature of 1000 °C
Lids are available separately.
Lids are available separately.
Certifications: According to DIN 12904.