Crucibles, tall form
Supplier: TECHNICAL QUARTZ SOLUTIONS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
73069 73073 73075 73077 73079
459-0238EA 141 GBP
459-0238 216-4490 216-4492 459-0180 459-0239
Crucibles, tall form
Crucibles
Fused quartz, translucent, glazed crucibles.
  • Max. temperature: 1050 °C
Ordering information: Crucible and lid have to be ordered separately.