Zeolite mordenite, ammonium

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

45877.14 45877.22 45877.36
45877.14EA 25 GBP
45877.14 45877.22 45877.36
Zeolite mordenite, ammonium
Zeolite mordenite, ammonium

CAS No.: 1318-02-1 Molecular Formula: SiO2:Al2O3 Physical Form: Powder, S.A. 500 m2/g, 20:1 mole ratio Appearance: White MDL No.: MFCD00132601


Applications: Zeolite mordenite is used for the acid-catalyzed isomerization of alkanes and aromatics

Formula: SiO₂:Al₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00132601
CAS Number: 1318-02-1
EINECS: 215-283-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR