Zeolite β, ammonium

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

45873.14 45873.22 45873.36
45873.14EA 25.2 GBP
45873.14 45873.22 45873.36
Zeolite β, ammonium
Zeolite beta, ammonium

CAS No.: 1318-02-1 Molecular Formula: SiO2:Al2O3 Physical Form: Powder, S.A. 680 m2/g, 25:1 mole ratio Appearance: White MDL No.: MFCD00132601


Applications: Zeolite beta is used in isomerization of waxes, and Friedel Crafts reactions (alkylation and acylation)

Formula: SiO₂:Al₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00132601
CAS Number: 1318-02-1
EINECS: 215-283-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR