L(-)-Bornyl acetate, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


MQ100

45855-1ML-F 45855-5ML-F
45855-1ML-FEA 93.3 GBP
45855-1ML-F 45855-5ML-F
L(-)-Bornyl acetate, Supelco®
L(-)-Bornyl acetate
Formula: C₁₂H₂₀O₂
MW: 196.29 g/mol
MDL Number: MFCD00867808
CAS Number: 5655-61-8
EINECS: 227-101-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75c5a88e265a2ea5d3dc91d7f58d72283bf5d706&vid=e0d7aed0ae6bab587c8d4964732470acf87d97eb&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR