Bis(tri-o-tolylphosphine)palladium(0) (14,9% Pd)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

45814.MD 45814.03
45814.MDEA 76.5 GBP
45814.MD 45814.03
Bis(tri-o-tolylphosphine)palladium(0) (14,9% Pd)
Bis(tri-o-tolylphosphine)palladium(0)

CAS No.: 69861-71-8 Molecular Formula: C42H42P2Pd Formula Weight: 715.16 Physical Form: Crystalline Sensitivity: Air Sensitive Appearance: Off-white to yellow Melting Point: 220-225° MDL No.: MFCD12911908

Formula: C₄₂H₄₂P₂Pd
MW: 715.16 g/mol
Melting Pt: 220…225 °C
Storage Temperature: Ambient
MDL Number: MFCD12911908
CAS Number: 69861-71-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR