VWR® ARISTAR®, Sulphate Standard Solution, 1000 mg/L SO₄²⁻ in Water (from K₂SO₄) Standard for Ion Chromatography
Supplier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
458062DEA 190 GBP
458062D 458064F
VWR® ARISTAR®, Sulphate Standard Solution, 1000 mg/L SO₄²⁻ in Water (from K₂SO₄) Standard for Ion Chromatography
Standards Anionic Standards Sulphate Standards
K₂SO₄ in H₂O.
Traceable to SRM from NIST, tested in an ISO Guide 34 / ISO17025 accredited laboratory
Supplied with certificate of analysis.