VWR®, Phosphate standard solutions, PO₄³⁻ in Water (from KH₂PO₄) standard for ion chromatography, ARISTAR®
Supplier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
458054DEA 219 GBP
458054D 458052B 458122V
VWR®, Phosphate standard solutions, PO₄³⁻ in Water (from KH₂PO₄) standard for ion chromatography, ARISTAR®
Standards Anionic Standards Phosphate Standards
PO₄ in H₂O.
Supplied with certificate of analysis.
Certifications: Traceable to SRM from NIST, tested in an ISO 17034 / ISO17025 accredited laboratory.