VWR® ARISTAR®, Tungsten Standard Solution, 10000 mg/L W in Water (from W), Standard for ICP
Supplier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
457172EEA 441 GBP
457172E 457174G
VWR® ARISTAR®, Tungsten Standard Solution, 10000 mg/L W in Water (from W), Standard for ICP
Standards Cationic Standards Tungsten Standards
W in H₂O tr.NH₄OH.
Traceable to SRM from NIST, tested in an ISO Guide 34 / ISO17025 accredited laboratory.
Supplied with certificate of analysis.